Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ (R)-(+)-SKF38393 Hydrochloride
Shop All MilliporeSigma Products
Click to view available options
:
5mg
Description
A benzazepine derivative that acts as a selective Dopamine D1/D5 receptor partial agonist. Examination of the isomers in several in vitro and in vivo tests for both central and peripheral dopaminergic activity revealed that activity resided almost exclusively in the R isomer. The results suggest that the properly oriented 1-phenyl substituent of 1 is important for dopamine-like activity. Reported to have stimulant and anorectic effects.
Specifications
Specifications
Form | Solid |
Content And Storage | Protect from light |
Quantity | 5 mg |
Color | White |
Description | 98% by HPLC |
Target Proteins | D1 |
Molecular Formula | C16H17NO2·HCl |
Solubility | H2O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction