Learn More
MilliporeSigma™ Calbiochem™ Ras/Rac Transformation Blocker, SCH 51344
Shop All MilliporeSigma ProductsSpecifications
Content And Storage
Protect from light
Form
Solid
Quantity
10 mg
Color
Light Yellow
Description
98% by HPLC
Target Proteins
Ras/Rac
CAS
171927-40-5
Molecular Formula
C16H20N4O3
Formula Weight
316.4g/mol
Solubility
DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.