Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ (S)-(-)-Raclopride

Catalog No. 5058770001 Shop All MilliporeSigma Products
Click to view available options
:
10mg

D2 -like dopamine receptor antagonist targeting D2 R (

D2-like dopamine receptor antagonist targeting D2R (pKi =8.0), D3R (pKi = 7.9), and 5-HT1A R (pKi=5.2). Used to explore D2 like receptor mediated effects in neuropathic pain, and in antagonism studies of DA receptors during heroin usage and cocaine. D2 Receptor selectivity in various preps shows IC50 values around 32 nM.

Specifications

Form Solid
Content And Storage Protect from light
Quantity 10 mg
Color Off-white
Description 99% by HPLC
Target Proteins D2
Molecular Formula C15H20Cl2N2O3
Solubility DMSO or EtOH
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.