Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SB-277011-A Dihydrochloride

Catalog No. 5054840001 Shop All MilliporeSigma Products
Click to view available options
:
5mg

A brain-penetrant, high-affinity, and selective dopamine D3 receptor antagonist

A brain-penetrant, high-affinity, and selective dopamine D3 receptor antagonist. High affinity for the hD3 receptor (pKi = 7.95) with 100-fold selectivity over the hD2 receptor and over 66 other receptors, enzymes, and ion channels. Rat data shows that the compound is selective for the D3 receptor over certain serotonergic and cholinergic receptors at doses up to 56.0mg/kg and strong physiological response at 32.0mg/kg.

Specifications

Quantity 5 mg
Form Solid
Content And Storage Hygroscopic, Protect from light
Color White
Description 98% by HPLC
Target Proteins D3
Molecular Formula C28H30N4O·2HCl
Solubility H2O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.