Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ SCH 23390 Hydrochloride
Shop All MilliporeSigma Products
Click to view available options
:
10mg
Description
A highly potent and selective dopamine D1-like receptor antagonist with a Ki of 0.2 and 0.3nM for the D1 and D5 dopamine receptor subtypes, respectively. Also an agonist at 5-HT1C and 5-HT2C receptors in vitro (Ki values are 6.3nM and 9.3nM respectively).
Specifications
Specifications
Quantity | 10 mg |
Content And Storage | Protect from light |
Form | Solid |
Color | Off-white |
Description | 99% by Chiral HPLC |
Target Proteins | D1 and D5 |
Molecular Formula | C17H18ClNO·HCl |
Solubility | H2O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction