Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Sumanirole Maleate

Catalog No. 5058960001 Shop All MilliporeSigma Products
Click to view available options
:
5mg

An extremely selective dopamine D2 receptor agonist (Ki =

An extremely selective dopamine D2 receptor agonist (Ki = 9nM). Commonly used in Parkinson′s Disease models and to reproduce cocaine′s discriminative stimulus effects. Sumanirole has greater than 200-fold selectivity for the D2 receptor subtype versus the other dopamine receptor subtypes in radioligand binding assays. In cell-based assays, sumanirole is a fully efficacious agonist, with EC50 values between 17 and 75 nM. In animals, sumanirole elicits many physiological responses attributed to D2-like receptor function.

Specifications

Content And Storage Protect from light
Quantity 5 mg
Form Solid
Color Off-white
Description 98% by HPLC
Target Proteins D2
Molecular Formula C11H13N3O·C4H4O4
Solubility DMSO
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.