Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TLR1/TLR2 Agonist II, CU-T12-9

Catalog No. 5325830001 Shop All MilliporeSigma Products
Click to view available options
:
10mg

A diphenyl substituted imidazole based compound that directly and selectively targets TLR1/2 and induces their dimerization and activates TLR1 and 2 signaling leading to NF-κB and AP-1 activation ( K D = 182 and 478 nM, respectively; EC50

A diphenyl substituted imidazole based compound that directly and selectively targets TLR1/2 and induces their dimerization and activates TLR1 and 2 signaling leading to NF-κB and AP-1 activation ( K D = 182 and 478 nM, respectively; EC50 = 52.9nM for TLR2 in HEK-Blue cells over-expressing hTLR2). Does not affect the dimerization of TLR2/TLR6 and exhibits poor affinity towards TLR3, TLR4, TLR5, TLR7 and TLR8. Shown to compete with Pam3CSK4 (Cat. No. 506350; K i = 45.4nM) for binding to TLR1/2 interface and enhance heterodimerization. Up-regulates the expression of TLR1, TLR2, TNFα, IL-10, and iNOS in RAW 264.7 macrophages in a time-dependent manner. Shown to be non-toxic up to 100μM concentration.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Specifications

Content And Storage Protect from light
Form Powder
Quantity 10 mg
Color Yellow
Description 98% by HPLC
Target Proteins TLR1/TLR2
Molecular Formula C17H13F3N4O2
Solubility DMSO
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.