Learn More
Aat Bioquest Calbryte™ 520 AM | 2x50ug
SDP
Supplier:  Aat Bioquest 20650
Encompass_Preferred
The intracellular calcium flux assay is a widely used method in monitoring signal transduction pathways and high throughput screening of G protein"coupled receptors (GPCRs) and calcium channel targets. Followed by Fluo-3 being introduced in 1989, Fluo-4, Fluo-8 and Cal-520 were later developed with improved signal/background ratio, and became the widely used Ca2+ indicators for confocal microscopy, flow cytometry and high throughput screening applications. However, there are still a few severe problems with Fluo-4. For example, as for Fluo-3, in all most all the intracellular calcium assays with Fluo-4 AM, probenecid is required to prevent the cell-loaded Fluo-4 from leaking out of cells. The use of probenecid with Fluo-4-based calcium assays compromises the assay results since probenecid is well-documented to have a variety of complicated cellular effects. Calbryte™ 520, AM is a novel fluorescent and cell-permeable indicator for the measurement of intracellular calcium.
Catalog No. NC1424566
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.