Learn More
TARGETMOL CHEMICALS INC CALCITRIOL 5MG
Supplier:  TARGETMOL CHEMICALS INC T6316
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Calcitriol (125-Dihydroxyvitamin D3) is the physiologically active form of vitamin D. It is formed primarily in the kidney by enzymatic hydroxylation of 25-hydroxycholecalciferol (CALCIFEDIOL). Its production is stimulated by low blood calcium levels and parathyroid hormone. Calcitriol increases intestinal absorption of calcium and phosphorus and in concert with parathyroid hormone increases bone resorption. purity: 99%
Catalog No. 50-257-6533
missing translation for 'QuantityLeft'