Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium
SCI_ED

Catalog No. p-4416747 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Reagent Grade - Granular
Reagent Grade - Turnings
This item is not returnable. View return policy
This item is not returnable. View return policy

Reacts with water or moisture

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]

Specifications

Color Silver-White
Density 1.54g/mL at 20°C/4°C
Linear Formula Ca
Quantity 25 g
Formula Weight 40.08
Packaging Reagent Grade - Granular
Chemical Name or Material Calcium
ShelfLife 2 years
DOTInformation DOT Class 4.3, II, : Dangerous When Wet
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.