Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium-binding-protein-P22 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Calcium-binding-protein-P22 |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Calcium-binding-protein-P22 Polyclonal specifically detects Calcium-binding-protein-P22 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Calcium-binding-protein-P22 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Calcineurin B homolog, Calcineurin homologous protein, calcium binding protein P22, Calcium-binding protein CHP, calcium-binding protein p22, SLC9A1 binding protein, SLC9A1BP | |
CHP1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20 | |
Polyclonal | |
Rabbit | |
Wnt Signaling Pathway | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
11261 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GFSHSQITRLYSRFTSLDKGENGTLSREDFQRIPELAINPLGDRIINAFFPEGEDQVNFRGFMRTLAHFRPIEDNEKSKDVNGPEPLNSRSNKLHFAFRLYDLDKDEKISRDELLQVLRMMVGVNIS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title