Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium Carbonate
SCI_ED

Catalog No. S25220B Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Natural Chalk - Solid
Reagent Grade - Solid
CO2 generation

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Color White
Quantity 500 g
Assay Percent Range 100% w/w
Linear Formula CaCO3
Formula Weight 100.09
Packaging Natural Chalk - Solid
Density ∽2.7 to 2.9g/mL
Chemical Name or Material Calcium Carbonate
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.