Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, 2.75% (w/v), Ricca Chemical
SDP

Catalog No. p-3251459 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 10035-04-8, 7732-18-5

Specifications

Color Colorless
CAS Min % 95.0, 3.45
CAS Max % 98.0, 3.59
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 2.75% (w/v)
Chemical Name or Material Calcium Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.