Learn More
Quality Biological Inc Calcium Chloride 2 M, Pack of 4 x 100ML
SDP
Supplier:  Quality Biological Inc 351130721
Encompass_Preferred
Calcium Chloride 2 M is useful in the preparation of competent E.coli and transformation of bacteria.
Catalog No. 50-983-268
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.