Learn More
Research Products International Corp Calcium Gluconate, Monohydrate, 250 Grams
SDP
Supplier:  Research Products International Corp C30050250.0
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Additional calcium (Ca2+) source in plant tissue culture media.
Catalog No. 50-213-227