Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
calcium homeostasis modulator 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | calcium homeostasis modulator 2 |
---|---|
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
calcium homeostasis modulator 2 Polyclonal antibody specifically detects calcium homeostasis modulator 2 in Human samples. It is validated for Western BlotSpecifications
calcium homeostasis modulator 2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology | |
PBS (pH 7.0) | |
51063 | |
IgG | |
Antigen and protein A Affinity-purified |
Western Blot 1:500-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
calcium homeostasis modulator 2, calcium homeostasis modulator protein 2, FAM26Bfamily with sequence similarity 26, member B, Protein FAM26B | |
Produced in rabbits immunized with E. coli-derived Human calcium homeostasis modulator 2 fragment. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title