Learn More
(-)-Calcium hydroxycitrate tribasic, ≥97.0% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Packaging
Bottomless Glass Bottle with Inserted Fused Cone
Quantity
10 mg
CAS
921226-01-9
Type
Analytical Standard
Linear Formula
C12H10 Ca3O16
Synonym
(-)-Hydroxycitric acid tricalcium salt
Formula Weight
530.43
Percent Purity
≥97.0% (HPLC)
Grade
Analytical Standard
Physical Form
Neat
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.