Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Methacrylate Hydrate, TCI America™
SDP

Catalog No. M054825G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 16809-88-4
Molecular Formula C8H12CaO4
Molecular Weight (g/mol) 212.258
MDL Number MFCD00151661
InChI Key QYOYGGOEWRXYDF-UHFFFAOYSA-N
Synonym Methacrylic Acid Calcium Salt
PubChem CID 131874840
IUPAC Name calcium;2-methylprop-2-enoic acid
SMILES CC(=C)C(=O)O.CC(=C)C(=O)O.[Ca]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 210.24
Physical Form Crystalline Powder
Chemical Name or Material Calcium Methacrylate Hydrate
EINECSNumber (2)-1027
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.