Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phosphate Dibasic, MP Biomedicals™

Catalog No. p-7139876 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100g
500g
1 kg

Calcium Phosphate Dibasic, hite Powder

  • It is used in animal feeds, as a mineral supplement, as an antacid, in the manufacturing of glass, in dental products and in fertilizers.
  • It has been used as a dietary supplement, and as an antacid.

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
Synonym Calcium dihydrogenphosphate, Acid calcium phosphate, Calcium biphosphate, Monocalcium orthophosphate
IUPAC Name calcium hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O

Specifications

Density ca. 2.3g/cm3 at 20°C (Lit.)
Color White
Melting Point ∼370°C with decomposition (literature)
Boiling Point Does not boil (Literature)
Quantity 100g
Solubility Information Soluble in 1 M Hydrochloric acid (100mg/mL - slightly hazy, colorless to slight yellowish-green solution).
Physical Form Powder
Assay 0.9829
storageNote1 Store at Room Temperature (15-30°C).
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.