Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Silicate, NF, Spectrum™ Chemical
SDP

Catalog No. p-7157188 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 g
1 kg
8 kg
15 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

C1643, 1344-95-2, CaO•SiO2

Spectrum™ Chemical Calcium Silicate, NF , also known as calcium orthosilicate, is used as an anticaking agent in pharmaceutical preparations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1344-95-2
Molecular Formula CaO3Si
Molecular Weight (g/mol) 116.16
InChI Key OYACROKNLOSFPA-UHFFFAOYSA-N
IUPAC Name calcium oxosilanebis(olate)
SMILES [Ca++].[O-][Si]([O-])=O

Specifications

CAS Max % 100%
pH 8.4-11.2
Quantity 1 kg
Grade NF
Loss on Ignition 20%
Packaging Poly Pail
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.