Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Standard, 10,000μg/mL in water with dilute nitric acid, Ultra Scientific

Catalog No. USICP120
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Click to view available options
Quantity:

Chemical Identifiers

CAS

10043-52-4, 7697-37-2, 7732-18-5

Molecular Formula

CaCl2

Molecular Weight (g/mol)

110.98

InChI Key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

PubChem CID

5284359

ChEBI

CHEBI:3312

IUPAC Name

calcium dichloride

SMILES

[Cl-].[Cl-].[Ca++]

Specifications

Quantity

125 mL

CAS Max %

94.0, 5.0, 1.0

Concentration or Composition (by Analyte or Components)

10, 000μg/mL

Analyte or Component Names

Calcium,water,nitric acid

Solvent or Matrix

Water with dilute nitric acid

Chemical Name or Material

Calcium Standard

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.