Learn More
Cpi International CALCIUM STANDARD
SDP
Supplier:  Cpi International S4400100091
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
CPI International's Peak Performance Calcium (Ca) standard is provided at a concentration of 1,000 ppm and a volume of 100 mL. 1,000 ug/mL. In 2% HNO3 Matrix - 18 month shelf-life.
Catalog No. NC0994821
missing translation for 'QuantityLeft'