Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Standard 5ppm/H2O, Reagents

Catalog No. 01335685
Click to view available options
Quantity:

Calcium Standard 5ppm/H2O, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material

Calcium Standard 5ppm/H2O

Packaging

Poly Bottle

CAS

7732-18-5

Quantity

1 L

Color

Colorless

For Use With (Equipment)

Water Analyzer

Grade

Reagent

Boiling Point

100°C

Specific Gravity

1.0001

For Use With (Application)

Water Analysis

Solubility

Completely miscible

Physical Form

Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.