Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium Sulfate, Dihydrate
SCI_ED

Catalog No. p-4416357 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade - Powder
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10101-41-4, 7778-16-9

Specifications

Color White
Quantity 500 g
Assay Percent Range >95% w/w
Molecular Formula CaH4O6S
Linear Formula CaSO4·2H2O
MDL Number MFCD00149625
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O
IUPAC Name calcium dihydrate sulfate
Molecular Weight (g/mol) 172.16
PubChem CID 24928
ChEBI CHEBI:32583
Formula Weight 172.17
Grade Reagent
Packaging Reagent Grade - Powder
Density 2.32g/mL
Chemical Name or Material Calcium Sulfate, Dihydrate
Show More Show Less
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.