Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium Sulfate, Hemihydrate
SCI_ED

Catalog No. S25231A Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Solid
This item is not returnable. View return policy
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10034-76-1
Molecular Formula Ca2H2O9S2
Molecular Weight (g/mol) 290.283
InChI Key ZOMBKNNSYQHRCA-UHFFFAOYSA-J
Synonym calcium sulfate hemihydrate, gypsum hemihydrate, calcium sulphate hemihydrate, unii-3rw091j48v, crystacal, densite, hemihydrate gypsum, densite gypsum, gipshalbhydrat, p 23 sulfate
PubChem CID 3033839
ChEBI CHEBI:32584
IUPAC Name dicalcium;disulfate;hydrate
SMILES O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ca+2].[Ca+2]

Specifications

Color Red
Quantity 2.5 kg
Assay Percent Range 100% w/v
Linear Formula CaSO4·1/2H2O
Formula Weight 145.15
Packaging Solid
Density 2.964g/mL
Chemical Name or Material Calcium Sulfate, Hemihydrate
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.