Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Thioglycolate Trihydrate 94.0+%, TCI America™
SDP

Catalog No. p-7136486 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 5793-98-6
Molecular Formula C4H14CaO7S2
Molecular Weight (g/mol) 278.347
MDL Number MFCD00043437
InChI Key YKVKKJPPSKLAFH-UHFFFAOYSA-N
Synonym calciumthioglycolatetrihydrate
PubChem CID 131881455
IUPAC Name calcium;2-sulfanylacetic acid;trihydrate
SMILES C(C(=O)O)S.C(C(=O)O)S.O.O.O.[Ca]

Specifications

Color White
Quantity 25 g
Formula Weight 130.17
Percent Purity ≥94.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium Thioglycolate Trihydrate
EINECSNumber (2)-1363
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.