Learn More
Inorganic Ventures 200.7WW CAL SOLUTION 1A 125mL (Inorganic Ventures)
SDP
Supplier:  Inorganic Ventures WWCAL1A125ML
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Dilution: 1:100, Analyte: Ag 50 ug/mL, Ba, B, Sr 100 ug/mL each, Cd, Cu, Mn 200 ug/mL each, Se 500 ug/mL, As and Mn 1000 ug/mL each.
Catalog No. 50-723-855
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.