Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Calibrex™ solutae 530 Bottle Top Dispenser

Fits most laboratory bottles either directly or using supplied adapters. Calibrex™ solutae 530 Bottle Top Dispensers feature excellent chemical resistance, high volumetry performance and simplified maintenance. Volume range stretches between 0.1 and

Supplier:  DWK Life Sciences W845016

 View more versions of this product

Catalog No. 03-410-908


Add to Cart

Description

Description

The Calibrex™ 530 solutae has a PFA-coated plunger preventing the plunger and barrel from seizing together. This enables trouble free distribution of salt solutions, weak and strong acids, as well as bases. The materials in contact with liquid flow are chemically inert, providing for stability and long instrument life.

  • Permanent fluid path visibility
  • Integrated calibration system
  • Choice of adjustment button with each unit
  • Long lasting performance stability
  • Autoclavable at 121°C / 250°F fully assembled
  • Two year warranty
Specifications

Specifications

Autoclavable
0.1 to 1 mL
PFA-coated plunger and integrated calibration
2 Years
Bottletop Dispenser
Fits most laboratory bottles directly or with supplied adapters
Calibrex solutae 530
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.