Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Camphor
SCI_ED

Catalog No. S25232A Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:
This item is not returnable. View return policy
This item is not returnable. View return policy
Kill and preserve moths

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS

76-22-2

Molecular Formula

C10H16O

Molecular Weight (g/mol)

152.24

MDL Number

MFCD00074738,MFCD00064149

InChI Key

DSSYKIVIOFKYAU-UHFFFAOYNA-N

Synonym

camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon

PubChem CID

2537

ChEBI

CHEBI:36773

IUPAC Name

1,7,7-trimethylbicyclo[2.2.1]heptan-2-one

SMILES

CC1(C)C2CCC1(C)C(=O)C2

Specifications

Color

White

Density

0.992g/mL at 25°C

Assay Percent Range

>96% w/v

Packaging

Reagent Grade

Linear Formula

C10H16O

Quantity

100 g

Formula Weight

152.25

Chemical Name or Material

Camphor

ShelfLife

5 years

DOTInformation

DOT Class 4.1, III, : Flammable Solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.