Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Camphor 98.0+%, TCI America™
SDP

Catalog No. C12515G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 464-48-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00064148,MFCD00074738,MFCD00064149,MFCD00064149
InChI Key DSSYKIVIOFKYAU-OMNKOJBGSA-N
Synonym --camphor, l-camphor, 1s---camphor, 1s,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, --bornan-2-one, s-camphor, l--camphor, 1s,4s-camphor, camphor,-, --alcanfor
PubChem CID 444294
ChEBI CHEBI:15397
IUPAC Name (1S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
SMILES CC1(C)C2CC[C@]1(C)C(=O)C2

Specifications

Melting Point 177°C
Color White
Quantity 5 g
UN Number 2717
Formula Weight 152.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Camphor
EINECSNumber (4)-0308&(4)-0601
RTECSNumber EX1250000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.