Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Candesartan Cilexetil 98.0+%, TCI America™
SDP

Catalog No. p-7125570 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 145040-37-5
Molecular Formula C33H34N6O6
Molecular Weight (g/mol) 610.671
MDL Number MFCD00871371
InChI Key GHOSNRCGJFBJIB-UHFFFAOYSA-N
Synonym candesartan cilexetil, atacand, amias, parapres, kenzen, candesartan cilexetil usan, candesartan cilexitil, candesartan cilextil, chembl1014
PubChem CID 2540
ChEBI CHEBI:3348
IUPAC Name 1-cyclohexyloxycarbonyloxyethyl 2-ethoxy-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
SMILES CCOC1=NC2=CC=CC(=C2N1CC3=CC=C(C=C3)C4=CC=CC=C4C5=NNN=N5)C(=O)OC(C)OC(=O)OC6CCCCC6

Specifications

Melting Point 159°C
Color White
Quantity 200 mg
Formula Weight 610.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Candesartan Cilexetil
RTECSNumber DD6672500
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.