Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canrenone, 97%, Spectrum™ Chemical
SDP

Catalog No. p-7157143 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle

C1274, 976-71-6, C22H28O3

Spectrum™ Chemical Canrenone , an aldosterone antagonist, can be used as a diuretic. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 976-71-6
Molecular Formula C22H28O3
Molecular Weight (g/mol) 340.46
InChI Key UJVLDDZCTMKXJK-UHFFFAOYNA-N
IUPAC Name 9a,11a-dimethyl-2,3,3a,3b,7,8,9,9a,9b,10,11,11a-dodecahydrospiro[cyclopenta[a]phenanthrene-1,2'-oxolane]-5',7-dione
SMILES CC12CCC3C(C=CC4=CC(=O)CCC34C)C1CCC21CCC(=O)O1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 97%
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.