Learn More
MTC Bio Screw cap for MTC Bio 2.0mL microtube, natural color, non-sterile, w/ installed O-ring, 1000/bag
Supplier:  MTC Bio C3175N
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
The newest addition to the ClearSeal™ line is made of 100% natural/colorless polypropylene with no markings, dyes or metallic content and they conform with FDA 21CFR. In response to popular demand for diagnostic sampling and reagent packaging applications, color coded caps are also available. The silicone O-ring ensures a leakproof seal throughout the temperature range of -90°C to +121° C. All ClearSeal™ tubes are RNase, DNase, pyrogen/endotoxin free and may be centrifuged at up to 22,000xg. Dimensions 13.1 x 46.8cm.
Catalog No. 50-226-8462