Learn More
MTC Bio Cap inserts for MTC Bio cryogenic vials, red, 500/pk
Supplier:  MTC Bio V5809R
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
MTC Bio cryogenic vials are designed to hold, store and preserve biological materials for days, weeks, years, or decades. These thick-walled tubes are outfitted with silicone O-rings that allow them to maintain their seal at temperatures as low as -196°C. Sterilized by gamma irradiation, the tubes are packed in reasealable bags of 50 tubes (10 bags per case) and are sterilzied by gamma. The self-standing starfoot base mates perfectly with a FREE included rack so that one hand is all that is need to install and remove caps. RNase, DNase, pyrogen, & endotoxin free
Catalog No. 50-226-8516