Learn More
MilliporeSigma™ Supelco™ Capillary Cleaving™ Tools and Accessories
Increases column performance by making the properly sized, squarely-cut ferrule to seal capillary column ends
Supplier: MilliporeSigma™ Supelco™ 23815
Description
- Makes scalpel-like cuts to polyimide-coated fused silica
- Leaves no jagged edges to create problems
- Industrial sapphire cutting edges remains sharp indefinitely
- Spring-loaded retractable blade reduces the chance of breakage if the tool is dropped
- Replacement sapphire cutting blades are available
Specifications
Making Scalpel-Like Cuts to Polyimide-Coated Fused Silica | |
Retractable Blade Version (#23814) | |
Sapphire Cutting Blade | |
Replacement Blade |
Cleaving is a trademark of Sigma-Aldrich Co. LLC