Learn More
Thermo Scientific™ Capitol Vial Hair/Nail Specimen Collection Kit
Catalog No. p-7114725 Shop All Thermo Scientific Products
Click to view available options
Description:
Thermo Scientific™ Capitol Vial Hair/Nail Specimen Collection Kits enable convenient collection and transport of samples for drugs of abuse or forensic testing.
Thermo Scientific™ Capitol Vial Hair/Nail Specimen Collection Kits enable convenient collection and transport of samples for drugs of abuse or forensic testing. Each kit contains a collection foil, transport envelope, specimen bag, and tamper-evident tape.
Features:
  • Tamper-evident closures
  • Collection containers are Class 10,000 aseptic products
  • Customized kits and OEM parts available
  • Leak-resistant & air-tight, with a lock seal
  • Twin valve design provides tighter seal
Specifications
Description
Absorbent 12 in. x 12 in. Cut