Learn More
ACROBiosystems resDetect™ HIV-1 p24 ELISA Kit (Residue Testing)
Supplier:  ACROBiosystems RESA05096TESTS
Encompass_Preferred
HIV-1 p24 ELISA Kit is based on ELISA sandwich method and designed to measure HIV-1 p24 levels in samples from CAR-T product preparation processing. It contains recombinant HIV-1 p24 and a pair of antibodies against the recombinant factor, which are provided by ACROBiosystems. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of HIV-1 p24 concentrations.
Catalog No. 50-271-8979
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.