Learn More
UIC Incorporated CARBON ANODE SOL
SDP
Supplier:  UIC Incorporated CM300002
Encompass
Carbon Anode Solution; Clear or light yellow liquid; B.P. 370 deg.F; Specific Gravity: 1.1; Stored in a cool, dry, well-ventilated location far from heat or ignition sources; Isolate from oxidizing agents
Catalog No. NC9028568
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.