Learn More
UIC Incorporated CARBON CATHODE SOLN
SDP
Supplier:  UIC Incorporated CM300001
Encompass
Carbon Cathode Solution; Clear or light yellow when acidic, blue when basic; B.P. 140 deg. to 190 deg.C; Specific Gravity: 1.09; Stored in a cool, dry, well-ventilated location far from heat or ignition sources
Catalog No. NC9925766
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.