Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbon Dioxide (Pure Gas), 99.8% Analytical Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
:
48 L
Specifications
Specifications
Quantity | 48 L |
Packaging | Cylinder |
Physical Form | Gas |
Molecular Formula | CO2 |
Linear Formula | CO2 |
MDL Number | MFCD00011491 |
UN Number | UN 1013 2.2 |
Formula Weight | 44.01 |
Percent Purity | 99.8% |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction