Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | -111.0°C |
Density | 1.2620g/mL |
Boiling Point | 46.0°C |
Quantity | 100 mL |
Flash Point | −30°C |
Packaging | Glass Bottle |
Refractive Index | 1.627 |
Assay Percent Range | 99.85% min. (GC) |
Specific Gravity | 1.262 |
Linear Formula | CS2 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. Causes damage to organs through prolonged or repeated exposure. Suspected of damaging fertility. Suspected of damaging the unborn child. Highly fla |
Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. |
EINECSNumber | 200-843-6 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction