Learn More
Sigma Aldrich Fine Chemicals Biosciences Carbon nanotube, single-walled (6,5) chirality, >=95% carbon basis (>=95% as carbon nanotubes), 0.78 nm average diameter | 308068-56-6 | 1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 7737351G
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Carbon nanotube, single-walled (6,5) chirality, >=95% carbon basis (>=95% as carbon nanotubes), 0.78 nm average diameter | Purity: >=95% carbon basis (>=95% as carbon nanotubes) | 308068-56-6 | 1G
Catalog No. 50-186-6040
missing translation for 'QuantityLeft'