Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Carbon Steel Magnetic Needle
SCI_ED

Catalog No. p-7203229 Shop All Eisco Products

Carbon steel magnetic needle is ideal for magnetism experiments in physics classrooms

  • Single, magnetic needle
  • Features bearing options for pivoting
  • Made of carbon steel
  • Needle Length: 4 in. (100 mm)
  • Great tool for physics classrooms

Specifications

Product Type Magnetic Needle
Grade 7 to Undergraduate
For Use With (Application) Physics Demonstrations and Experiments
Includes Brass Cup Bearing
Material Carbon Steel
Quantity 1 Ea.
Class Size 1 Lab Group
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.