Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon tetrachloride, ≥99.95% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187431 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL
Carbon tetrachloride (CCl4) is a clear, colourless, volatile and very stable chlorinated hydrocarbon. It is widely used as a solvent for fats, varnishes, oils, lacquers, resins, and rubber waxes. It is also used as a dry-cleaning agent, as a refrigerant, and as a starting material in the manufacturing of organic compounds. Featured Industries: Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 56-23-5
Quantity 1 mL
Type Analytical Standard
Linear Formula CCl4
MDL Number MFCD00000785
Synonym Tetrachloromethane
Formula Weight 153.82
Percent Purity ≥99.95% (GC)
Grade Analytical Standard
Vapor Pressure 143 mmHg (30°C)
Vapor Density 5.32 (vs. air)
Density 1.594 g/mL (at 25°C)
Melting Point -23°C
Boiling Point 76°C to 77°C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.