Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Tetrachloride, 99%, Thermo Scientific™

Catalog No. p-2671736
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 56-23-5
Molecular Formula CCl4
Molecular Weight (g/mol) 153.81
MDL Number MFCD00000785
InChI Key VZGDMQKNWNREIO-UHFFFAOYSA-N
Synonym carbon tetrachloride, perchloromethane, tetrasol, methane, tetrachloro, vermoestricid, benzinoform, necatorine, tetrafinol, carbona, univerm
PubChem CID 5943
ChEBI CHEBI:27385
IUPAC Name tetrachloromethane
SMILES ClC(Cl)(Cl)Cl

Specifications

Decomposition Information Decomposes information: >100°C
Melting Point -23°C
Density 1.594g/mL
Boiling Point 76°C (760mm Hg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4590 to 1.4610 (20°C, 589nm)
Linear Formula CCl4
Quantity 1 L
UN Number 1846
Residue after Evaporation 0.001% max.
Beilstein 01, 64
Fieser 03,43; 04,69; 12,551; 14,72; 16,367
Merck Index 15, 1816
Formula Weight 153.82
Percent Purity ≥98.5%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Carbon Tetrachloride
Show More Show Less
Hazard Category Signal Word: DANGER!
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Toxic in contact with skin.
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Toxic if swallowed.
Harms public health and the environment by de
Precautionary Statement Emergency Overview
Toxic by inhalation, in contact with skin and if swallowed. Toxic: danger of serious damage to health by prolonged exposure through inhalation. Suspected human carcinogen. May cause eye, skin, and respiratory tract irritation . May cause central nervous system effects. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Harms public health and the environment by destroying ozone in the upper atmosphere.
NFPA
Health:3 Flammability:0 Instability:0
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.