Learn More
Carolina™ Brine Shrimp Eggs

Shop All Carolina Biological Supply Products


- Brine shrimp eggs hatch in about two days when put in a weak salt solution
- Excellent food for hydra and small aquarium fish
- With instructions
Specifications
Packaging
Vial
Storage Requirements
26°C to 28°C
Quantity
1 oz.
Scientific Name
Artemia
Includes
Instructions
Sufficient For
Class of 30 students
Product Type
Brine Shrimp Egg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.