Learn More
Carolina™ Aquaspirillum itersonii, Living, Tube
Shop All Carolina Biological Supply Products
Click to view available options
:
1 Tube
Description
It is cultured on nutrient agar at 25 to 30 degrees C in a tube. Cultures are guaranteed as to species and purity.
- Each culture contains enough material for a class of 30 students.
Specifications
Specifications
Content And Storage | 25 to 30 °C |
Gram Stain Result | Gram (-) |
Media Recommendation | Nutrient Agar |
Product Type | Bacterial Culture |
Quantity | 1 Tube |