Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Streptococcus agalactiae, MicroKwik Culture™, PATHOGEN, Vial
SDPSCI_ED

Catalog No. S21825ND Shop All Carolina Biological Supply Products
Click to view available options
:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

Note: Each MicroKwik Culture™ comes with 5mL of rehydration medium.

This Biosafety Level 2 (pathogen) culture is freeze-dried cells plus medium.

Every culture is guaranteed for species and purity. Bacteria cultures are labeled by genus and species name, media on which they are cultivated, and temperature for optimal growth. All Gram staining is based on 24-hour cultures. Special packing and transportation costs are extra for animal and human pathogens.

  • Each culture contains enough material for a class of 30 students
  • Gram+, nonmotile cocci in pairs
  • Cultured on tryptic soy agar with 5% sheep blood at 37°C

Specifications

Gram Stain Result

Gram+

Media Recommendation

tryptic soy agar with 5% sheep blood

Product Type

Bacterial Culture

Quantity

1 vial

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.