Learn More
Carolina™ Individual Ciliate Cultures
SDPSCI_ED
Most ciliates are heterotrophs, feeding on smaller organisms, such as bacteria and algae.
Supplier:  Carolina Biological Supply 131620
 View more versions of this product
Catalog No. S07456ND
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
Description
Note: To ensure you order enough living materials for the number of students in your class, order 1 culture for up to 30 students, 2 cultures for 31 to 60 students, 3 cultures for 61 to 90 students, etc.
Specifications
Specifications
Tube
Ea.
Dropping pipet, appropriate medium
Tetrahymena Medium
22°C
20°C to 22°C
Tetrahymena
Class of 30 students
Tetrahymena
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.