Learn More
Carolina™ Individual Green Algae Cultures
SDPSCI_ED
Green algae are eukaryotic organisms that follow a reproduction cycle called alternation of generations.
Supplier:  Carolina Biological Supply 152665
 View more versions of this product
Catalog No. S07488ND
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
Description
Low light is 50 to 100 ft. candles of fluorescent light or fluorescent light from the ceiling; High light level is 200 to 400 ft. candles of fluorescent light 18 to 24 inches from the culture.
Specifications
Specifications
Ea.
Carolina™ culturing algae manual
Alga-Gro™ freshwater or soil water
22°C
V. globator
Class of 30 students
Volvox globator
High
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.